siRNA Details
Gene Symbol: Pcdh15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142735.1 | NP_001136207.1 | 3 | 550 | ||
NM_001142736.1 | NP_001136208.1 | 3 | 550 | |||
NM_001142737.1 | NP_001136209.1 | 3 | 550 | |||
NM_001142738.1 | NP_001136210.1 | 3 | 550 | |||
NM_001142740.1 | NP_001136212.1 | 3 | 550 | |||
NM_001142742.1 | NP_001136214.1 | 4 | 565 | |||
NM_001142743.1 | NP_001136215.1 | 3 | 550 | |||
NM_001142746.1 | NP_001136218.1 | 4 | 565 | |||
NM_001142747.1 | NP_001136219.1 | 4 | 565 | |||
NM_001142748.1 | NP_001136220.1 | 3 | 550 | |||
NM_001142760.1 | NP_001136232.1 | 3 | 550 | |||
NM_023115.3 | NP_075604.2 | 4 | 565 | |||
GenBank mRNA | AF281899.1 | - | 4 | 549 | ||
AK139154.1 | - | 4 | 490 | |||
DQ354396.1 | - | 3 | 534 | |||
DQ354397.1 | - | 3 | 534 | |||
DQ354398.1 | - | 3 | 534 | |||
DQ354399.1 | - | 3 | 534 | |||
DQ354400.1 | - | 3 | 534 | |||
DQ354401.1 | - | 3 | 534 | |||
DQ354403.1 | - | 3 | 534 | |||
DQ354405.1 | - | 4 | 549 | |||
DQ354406.1 | - | 3 | 534 | |||
DQ354408.1 | - | 3 | 534 | |||
DQ354409.1 | - | 3 | 534 | |||
DQ354410.1 | - | 3 | 534 | |||
DQ354411.1 | - | 3 | 534 | |||
DQ354412.1 | - | 3 | 534 | |||
DQ354413.1 | - | 4 | 549 | |||
DQ354414.1 | - | 3 | 534 | |||
DQ354416.1 | - | 4 | 490 | |||
DQ354417.1 | - | 3 | 475 | |||
DQ354418.1 | - | 3 | 475 |
