siRNA Details
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023118.5 | NP_075607.2 | 10 | 1302 | ||
GenBank mRNA | AK154716.1 | - | 10 | 1286 | ||
CB598046.1 | - | 1 | 343 |

More Information
Other Species Targeted

Gene Ontology Categories:
