siRNA Details
Gene Symbol: Cys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138686.3 | NP_619627.3 | 4, 5 | 1431 | ||
GenBank mRNA | AF390548.1 | - | 4, 5 | 1449 | ||
AK013490.1 | - | 4, 5 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
