siRNA Details
Gene Symbol: Entpd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172117.5 | NP_742115.2 | 5, 6 | 782 | ||
GenBank mRNA | AK030789.1 | - | 5, 6 | 783 | ||
BC038126.1 | - | 5, 6 | 771 | 1348160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
