siRNA Details
Gene Symbol: Camk2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039138.2 | NP_001034227.1 | 20 | 2372 | ||
NM_001039139.2 | NP_001034228.1 | 19 | 2303 | |||
NM_178597.5 | NP_848712.2 | 21 | 2405 | |||
GenBank mRNA | AK031213.1 | - | 19 | 2266 | ||
AK078311.1 | - | 20 | 2318 | |||
AK154764.1 | - | 20 | 2302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
