siRNA Details
Gene Symbol: Camk2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039138.2 | NP_001034227.1 | 20 | 1942 | ||
NM_001039139.2 | NP_001034228.1 | 19 | 1873 | |||
NM_178597.5 | NP_848712.2 | 21 | 1975 | |||
GenBank mRNA | AK031213.1 | - | 19 | 1836 | ||
AK078311.1 | - | 20 | 1888 | |||
BC025597.1 | - | 19 | 1827 | 5342641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
