siRNA Details
Gene Symbol: Adnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009628.2 | NP_033758.2 | 4 | 1486 | ||
GenBank mRNA | AF068198.1 | - | 1 | 152 | ||
AK129214.1 | - | 4 | 1537 | |||
AK134786.1 | - | 3 | 1008 | |||
BC050833.1 | - | 1 | 435 | |||
BC057666.1 | - | 1 | 544 | |||
BC090840.1 | - | 1 | 682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
