siRNA Details
Gene Symbol: Mrps33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010270.2 | NP_034400.1 | 2 | 277 | ||
GenBank mRNA | AK019000.1 | - | 2 | 229 | ||
AK144131.1 | - | 2 | 234 | |||
BC092135.1 | - | 1 | 181 | 5713592 | ||
BC096397.1 | - | 1 | 224 | 3466818 | ||
BU695761.1 | - | 2 | 356 | |||
BY296250.1 | - | 2 | 272 | |||
Y17852.1 | - | 2 | 243 |

More Information
Other Species Targeted

Gene Ontology Categories:
