siRNA Details
Gene Symbol: Add1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024458.3 | NP_001019629.2 | 4 | 622 | ||
NM_001102444.1 | NP_001095914.1 | 4 | 622 | |||
NM_013457.3 | NP_038485.1 | 4 | 622 | |||
GenBank mRNA | AF096839.1 | - | 4 | 520 | ||
AF189771.1 | - | 3 | 402 | |||
AK054558.1 | - | 4 | 474 | |||
AK079960.1 | - | 4 | 477 | |||
CD349465.1 | - | 4 | 622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
