siRNA Details
Gene Symbol: SMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002800.2 | NP_001002800.1 | 3 | 612 | ||
NM_005496.3 | NP_005487.3 | 2 | 429 | |||
XM_006713459.3 | XP_006713522.1 | 3 | 382 | |||
XM_011512311.2 | XP_011510613.1 | 3 | 500 | |||
XM_011512312.2 | XP_011510614.1 | 2 | 121 | |||
GenBank mRNA | AB019987.1 | - | 3 | 273 | ||
AK002200.1 | - | 3 | 266 | |||
AK225437.1 | - | 3 | 266 | |||
AK315070.1 | - | 3 | 266 | |||
AL136877.1 | - | 2 | 393 | |||
AL833949.1 | - | 2 | 1320 | |||
BC001557.1 | - | 3 | 248 | |||
BC065259.1 | - | 3 | 444 | |||
BC070161.1 | - | 3 | 266 | |||
BC106033.1 | - | 3 | 284 | |||
BI090399.1 | - | 287 | ||||
DB444342.2 | - | 3 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
