siRNA Details
Gene Symbol: Bcl2l14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025778.3 | NP_080054.1 | 4 | 787 | ||
GenBank mRNA | AK008682.1 | - | 3 | 712 | ||
AK016670.1 | - | 4 | 788 | |||
AK016997.1 | - | 4 | 787 | |||
AK018579.1 | - | 3 | 754 | |||
AK035707.1 | - | 3 | 719 | |||
AK160247.1 | - | 4 | 790 | |||
BC025541.1 | - | 3 | 707 | 5253808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
