siRNA Details
Gene Symbol: Bcl2l14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025778.3 | NP_080054.1 | 4 | 845 | ||
GenBank mRNA | AK008682.1 | - | 3 | 770 | ||
AK016670.1 | - | 4 | 846 | |||
AK016997.1 | - | 4 | 845 | |||
AK018579.1 | - | 3 | 812 | |||
AK035707.1 | - | 3 | 777 | |||
AK160247.1 | - | 4 | 848 |
