siRNA Details
Gene Symbol: ING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005537.5 | NP_005528.4 | 1 | 584 | ||
GenBank mRNA | AB024402.1 | - | 1 | 613 | ||
AB031269.1 | - | 1 | 28 | |||
AF181849.1 | - | 1 | 584 | |||
BC018348.1 | - | 1 | 19 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
