siRNA Details
Gene Symbol: Cfap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027559.2 | NP_081835.2 | 5 | 750 | ||
GenBank mRNA | AK014581.1 | - | 768 | |||
AK015217.1 | - | 5 | 772 | |||
AK015497.1 | - | 5 | 751 |

More Information
Other Species Targeted

Gene Ontology Categories:
