siRNA Details
Gene Symbol: KRTAP5-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021046.2 | NP_066384.2 | 1 | 270 | ||
GenBank mRNA | AB126077.1 | - | 1 | 270 | ||
AY360461.1 | - | 1 | 239 | |||
BC119736.1 | - | 3 | 323 |
Gene Symbol: KRTAP5-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005553.3 | NP_005544.4 | 1 | 453 | ||
GenBank mRNA | AB126078.1 | - | 1 | 249 | ||
BC069531.1 | - | 1 | 215 | 7262098 | ||
BC101744.1 | - | 1 | 357 | 8069250 | ||
X63755.1 | - | 1 | 453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
