siRNA Details
Gene Symbol: KRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005556.3 | NP_005547.3 | 2, 3 | 657 | ||
XM_011538325.1 | XP_011536627.1 | 2, 3 | 610 | |||
XM_017019294.1 | XP_016874783.1 | 2, 3 | 235 | |||
GenBank mRNA | AF509887.1 | - | 2, 3 | 586 | ||
AJ238246.1 | - | 2, 3 | 591 | |||
AK025881.1 | - | 2, 3 | 584 | |||
AK128505.1 | - | 2, 3 | 476 | |||
AK129924.1 | - | 3, 4 | 591 | |||
AK225498.1 | - | 2, 3 | 580 | |||
BC002700.2 | - | 2, 3 | 572 | 3610347 | ||
BE744321.1 | - | 234 | ||||
BU527162.1 | - | 2, 3 | 658 | |||
X03212.1 | - | 2, 3 | 591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
