siRNA Details
Gene Symbol: 4930503B20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029144.3 | NP_083420.2 | 1 | 357 | ||
GenBank mRNA | AK007081.1 | - | 1 | 360 | ||
AK015617.1 | - | 1 | 353 | |||
AK015685.1 | - | 1 | 355 | |||
BC048658.1 | - | 1 | 357 | 6773189 |
