siRNA Details
Gene Symbol: Ppp1r12c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029834.3 | NP_084110.2 | 2 | 441 | ||
GenBank mRNA | AK010836.1 | - | 2 | 427 | ||
AK046570.1 | - | 2 | 411 | |||
AK144521.1 | - | 2 | 385 | |||
AK144691.1 | - | 2 | 442 | |||
AK187068.1 | - | 2 | 52 |

More Information
Other Species Targeted

Gene Ontology Categories:
