siRNA Details
Gene Symbol: Zfp623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030199.3 | NP_084475.1 | 2 | 2430 | ||
GenBank mRNA | AK019163.1 | - | 2 | 2327 | ||
AK030476.1 | - | 2 | 2335 | |||
AK046806.1 | - | 2 | 2431 | |||
AK169386.1 | - | 2 | 2428 | |||
AK172985.1 | - | 2 | 2405 | |||
AK188875.1 | - | 1 | 237 | |||
AY610936.1 | - | 2 | 2431 | |||
BC086767.1 | - | 2 | 2321 | 6832277 |
