siRNA Details
Gene Symbol: Actn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033268.4 | NP_150371.4 | 21 | 2915 | ||
GenBank mRNA | AF248643.1 | - | 21 | 2683 | ||
AK003570.1 | - | 5 | 678 | |||
AY036877.1 | - | 22 | 2859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
