siRNA Details
Gene Symbol: Stim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081103.2 | NP_001074572.2 | 12 | 2914 | ||
GenBank mRNA | AK041804.1 | - | 6 | 1468 | ||
AK173183.1 | - | 12 | 2914 | |||
BC043455.1 | - | 5 | 1436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
