siRNA Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081346.1 | NP_001074815.1 | 13 | 2125 | ||
GenBank mRNA | AK030986.1 | - | 12 | 2006 | ||
AK045134.1 | - | 2022 | ||||
AK082859.1 | - | 5 | 1161 | |||
AK087079.1 | - | 3 | 870 | |||
AK087239.1 | - | 4 | 1035 | |||
AY036116.2 | - | 1 | 665 | |||
BC158093.1 | - | 12 | 2127 | 9007380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
