siRNA Details
Gene Symbol: PDLIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005451.4 | NP_005442.2 | 10 | 1009 | ||
NM_203352.2 | NP_976227.1 | 10 | 907 | |||
NR_103804.1 | 10 | 1208 | ||||
XM_011534698.2 | XP_011533000.1 | 9 | 886 | |||
XM_011534699.2 | XP_011533001.1 | 11 | 994 | |||
GenBank mRNA | AF265209.1 | - | 10 | 1004 | ||
AF345904.1 | - | 10 | 965 | |||
AF345905.1 | - | 10 | 863 | |||
AF345906.1 | - | 11 | 982 | |||
AK096826.1 | - | 8 | 1599 | |||
BC001093.2 | - | 10 | 987 | 3506748 | ||
BC067806.1 | - | 10 | 1204 | |||
BC084575.1 | - | 9 | 925 | 6729368 | ||
BU956109.1 | - | 3 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
