siRNA Details
Gene Symbol: GUCA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005459.3 | NP_005450.3 | 1, 2 | 280 | ||
XM_005247910.3 | XP_005247967.1 | 1, 2 | 356 | |||
GenBank mRNA | AF110003.1 | - | 1, 2 | 151 | ||
AV732101.1 | - | 1, 2 | 280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
