siRNA Details
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002751.6 | NP_002742.3 | 11, 12 | 1123 | ||
NR_110887.1 | 11, 12 | 1123 | ||||
XM_011530706.1 | XP_011529008.1 | 9, 10 | 847 | |||
GenBank mRNA | AF001008.1 | - | 11, 12 | 1022 | ||
AF001174.1 | - | 11, 12 | 1028 | |||
AF031135.1 | - | 11, 12 | 1037 | |||
AK291845.1 | - | 11, 12 | 1074 | |||
AK308969.1 | - | 10, 11 | 960 | |||
BC027933.1 | - | 11, 12 | 1080 | 5199628 | ||
CR456514.1 | - | 11, 12 | 1024 | |||
U53442.1 | - | 11, 12 | 1065 | |||
U92268.1 | - | 11, 12 | 1026 | |||
Y14440.1 | - | 11, 12 | 1028 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
