siRNA Details
Gene Symbol: ACOT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005469.3 | NP_005460.2 | 1, 2 | 232 | ||
XM_011528480.2 | XP_011526782.1 | 1, 2 | 194 | |||
GenBank mRNA | AF014404.1 | - | 1, 2 | 168 | ||
AF124264.1 | - | 1, 2 | 138 | |||
AK296973.1 | - | 1, 2 | 220 | |||
BC117155.1 | - | 1, 2 | 160 | 40125706 | ||
BC117157.1 | - | 1, 2 | 160 | 40125708 | ||
BI911038.1 | - | 1, 2 | 218 | |||
CR541771.1 | - | 1, 2 | 138 | |||
CR541778.1 | - | 1, 2 | 138 | |||
DB470803.3 | - | 1, 2 | 232 | |||
X86032.1 | - | 1, 2 | 206 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
