siRNA Details
Gene Symbol: Pxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK156146.1 | - | 3 | 246 | ||
AK157364.1 | - | 3 | 274 | |||
AK172347.1 | - | 3 | 229 | |||
AK172463.1 | - | 3 | 281 | |||
BC016131.1 | - | 3 | 250 | 4922274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
