siRNA Details
Gene Symbol: Pxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145458.3 | NP_663433.2 | 18 | 2463 | ||
NM_178279.2 | NP_840063.1 | 19 | 2496 | |||
GenBank mRNA | AK028501.1 | - | 18 | 2413 | ||
AK048910.1 | - | 18 | 2420 | |||
AK147037.1 | - | 18 | 2460 | |||
AK150337.1 | - | 18 | 2445 | |||
AK150413.1 | - | 18 | 2445 | |||
AK150660.1 | - | 18 | 2461 | |||
AK151098.1 | - | 18 | 2424 | |||
AK152058.1 | - | 18 | 2422 | |||
AK152789.1 | - | 18 | 2422 | |||
AK153067.1 | - | 18 | 2422 | |||
AK156146.1 | - | 19 | 2503 | |||
AK157364.1 | - | 18 | 2453 | |||
AK159541.1 | - | 6 | 1193 | |||
AK166036.1 | - | 18 | 2389 | |||
AK169112.1 | - | 18 | 2421 | |||
AK170361.1 | - | 18 | 2456 | |||
AK172463.1 | - | 18 | 2462 | |||
BC016131.1 | - | 18 | 2433 | 4922274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
