siRNA Details
Gene Symbol: Sfswap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172276.3 | NP_758480.2 | 5 | 802 | ||
GenBank mRNA | AK031872.1 | - | 5 | 793 | ||
AK088182.1 | - | 5 | 746 | |||
AK140369.1 | - | 5 | 795 | |||
BC057314.1 | - | 5 | 2732 |

More Information
Other Species Targeted

Gene Ontology Categories:
