siRNA Details
Gene Symbol: Aim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172393.2 | NP_765981.2 | 2 | 3558 | ||
GenBank mRNA | AK041082.1 | - | 1 | 174 | ||
AK088102.1 | - | 1 | 230 | |||
BC035551.1 | - | 1 | 710 | |||
BC059272.1 | - | 1 | 715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
