siRNA Details
Gene Symbol: Aim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172393.2 | NP_765981.2 | 18 | 5455 | ||
GenBank mRNA | AF397913.1 | - | 12 | 1163 | ||
AK041082.1 | - | 17 | 2072 | |||
AK088102.1 | - | 17 | 2127 | |||
BC031829.1 | - | 5 | 468 | |||
BC035551.1 | - | 17 | 2607 | |||
BC059272.1 | - | 17 | 2612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
