siRNA Details
Gene Symbol: Exog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172136.1 | NP_001165607.1 | 3 | 345 | ||
NM_172456.3 | NP_766044.1 | 2 | 310 | |||
GenBank mRNA | AK028887.1 | - | 2 | 274 | ||
AK034654.1 | - | 3 | 307 | |||
AK051300.1 | - | 3 | 309 | |||
AK162566.1 | - | 2 | 273 | |||
AK163555.1 | - | 2 | 274 | |||
BC052171.1 | - | 2 | 256 | |||
BC059920.1 | - | 3 | 288 | |||
BC064730.1 | - | 3 | 360 | |||
BC094589.1 | - | 2 | 257 | |||
BC119024.1 | - | 2 | 274 | 8733773 | ||
BY113415.1 | - | 2 | 311 |
