siRNA Details
Gene Symbol: Zswim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172503.3 | NP_766091.2 | 2 | 259 | ||
GenBank mRNA | AK041934.1 | - | 2 | 273 | ||
AK052168.1 | - | 2 | 273 | |||
AK053663.1 | - | 2 | 273 | |||
AK150221.1 | - | 2 | 273 | |||
AK155465.1 | - | 2 | 273 | |||
BC058080.1 | - | 2 | 259 | 6822376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
