siRNA Details
Gene Symbol: Map3k13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172821.3 | NP_766409.2 | 10 | 2184 | ||
GenBank mRNA | AK047986.1 | - | 1 | 258 | ||
AK051378.1 | - | 1 | 177 | |||
DQ480427.1 | - | 10 | 2184 |

More Information
Other Species Targeted

Gene Ontology Categories:
