siRNA Details
Gene Symbol: Map3k13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172821.3 | NP_766409.2 | 13 | 4829 | ||
GenBank mRNA | AK047986.1 | - | 4 | 2903 | ||
AK051378.1 | - | 4 | 2826 | |||
AK054039.1 | - | 1 | 1665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
