siRNA Details
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172988.4 | NP_766576.1 | 4 | 865 | ||
GenBank mRNA | AK028786.1 | - | 4 | 849 | ||
AK032391.1 | - | 4 | 844 | |||
AK051429.1 | - | 4 | 816 | |||
AK083661.1 | - | 4 | 839 | |||
BC033265.1 | - | 4 | 792 | 3983942 |
