siRNA Details
Gene Symbol: IDH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005530.2 | NP_005521.1 | 4 | 252 | ||
XM_017022118.1 | XP_016877607.1 | 3 | 267 | |||
GenBank mRNA | AK093531.1 | - | 2 | 760 | ||
AK222973.1 | - | 4 | 236 | |||
AK296002.1 | - | 4 | 227 | |||
AK299970.1 | - | 4 | 269 | |||
AL442090.1 | - | 1 | 174 | |||
BC021967.2 | - | 4 | 245 | 2989636 | ||
BC036227.1 | - | 4 | 253 | |||
CR749548.1 | - | 4 | 891 | |||
U07681.1 | - | 4 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
