siRNA Details
Gene Symbol: INPP5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005541.4 | NP_005532.2 | 3, 4 | 859 | ||
GenBank mRNA | AK309644.1 | - | 3, 4 | 484 | ||
BC099920.1 | - | 3, 4 | 470 | 6304992 | ||
BC113580.1 | - | 3, 4 | 407 | 8322632 | ||
BC113582.1 | - | 3, 4 | 407 | 8322634 | ||
U57650.1 | - | 3, 4 | 859 | |||
X98429.1 | - | 3, 4 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
