siRNA Details
Gene Symbol: Adrbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285806.1 | NP_001272735.1 | 13 | 1198 | ||
NM_177078.4 | NP_796052.2 | 13 | 1159 | |||
GenBank mRNA | AK076533.1 | - | 13 | 1057 | ||
AK134672.1 | - | 13 | 1136 | |||
AK141186.1 | - | 13 | 1178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
