siRNA Details
Gene Symbol: KLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002231.2 | NP_001002231.1 | 4 | 555 | ||
NM_001256080.1 | NP_001243009.1 | 3 | 395 | |||
NM_005551.4 | NP_005542.1 | 4 | 555 | |||
NR_045763.1 | 4 | 641 | ||||
GenBank mRNA | AF188746.1 | - | 4 | 530 | ||
AF188747.1 | - | 4 | 530 | |||
AK300527.1 | - | 4 | 641 | |||
AK300549.1 | - | 3 | 389 | |||
AK300566.1 | - | 4 | 549 | |||
AK309982.1 | - | 2 | 508 | |||
AK309985.1 | - | 3 | 392 | |||
AY429509.1 | - | 3 | 656 | |||
AY429510.1 | - | 2 | 496 | |||
BC005196.1 | - | 4 | 555 | 3951036 | ||
BT006650.1 | - | 4 | 514 | |||
S39329.1 | - | 4 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
