siRNA Details
Gene Symbol: Zfp236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177832.3 | NP_808500.2 | 30 | 6427 | ||
GenBank mRNA | AK041130.1 | - | 8 | 2060 | ||
AK044757.1 | - | 1 | 43 | |||
AK082879.1 | - | 7 | 2075 | |||
AK151217.1 | - | 1 | 1114 | |||
AK156842.1 | - | 10 | 2530 | |||
AK167703.1 | - | 4 | 1547 | |||
BC036962.1 | - | 6 | 1849 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
