siRNA Details
Gene Symbol: Teddm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178243.3 | NP_839974.2 | 1 | 705 | ||
GenBank mRNA | AJ554213.1 | - | 1 | 662 | ||
AK136655.1 | - | 1 | 707 | |||
AK136685.1 | - | 1 | 706 | |||
BC120491.1 | - | 1 | 659 | 40129414 | ||
BC131905.1 | - | 1 | 661 | 40130182 | ||
DQ402363.1 | - | 1 | 705 |
