siRNA Details
Gene Symbol: Eml5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081191.1 | NP_001074660.1 | 41 | 6024 | ||
GenBank mRNA | AK044024.1 | - | 5 | 677 | ||
BC027154.1 | - | 4 | 569 | |||
BC158137.1 | - | 39 | 5496 | 9007401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
