siRNA Details
Gene Symbol: Eml5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081191.1 | NP_001074660.1 | 44 | 6901 | ||
GenBank mRNA | AK044024.1 | - | 8 | 1554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
