siRNA Details
Gene Symbol: TRIM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003852.3 | NP_003843.3 | 19 | 3168 | ||
NM_015905.2 | NP_056989.2 | 19 | 3270 | |||
GenBank mRNA | AF009353.1 | - | 19 | 2941 | ||
AF119042.1 | - | 19 | 3069 | |||
AK075306.1 | - | 19 | 3270 | |||
AK302679.1 | - | 20 | 3143 | |||
AK312610.1 | - | 19 | 3296 | |||
BC028689.2 | - | 19 | 3168 | 4827559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
