siRNA Details
Gene Symbol: Zxdb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081473.1 | NP_001074942.1 | 1 | 3007 | ||
GenBank mRNA | AK016708.1 | - | 1 | 2095 | ||
AK132358.1 | - | 1 | 2150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
