siRNA Details
Gene Symbol: Zfp598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183149.1 | NP_898972.1 | 2 | 326 | ||
GenBank mRNA | AK131127.1 | - | 2 | 345 | ||
AK220149.1 | - | 1 | 19 | |||
BC024690.1 | - | 2 | 120 | |||
BC050859.1 | - | 2 | 326 | 6328878 |
