siRNA Details
Gene Symbol: Polr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081247.1 | NP_001074716.1 | 2 | 243 | ||
GenBank mRNA | AK039440.1 | - | 2 | 256 | ||
AK143315.1 | - | 2 | 193 | |||
AK170475.1 | - | 2 | 250 | |||
BC053071.1 | - | 2 | 206 | |||
BC151002.1 | - | 2 | 153 | 9087915 | ||
BC151168.1 | - | 2 | 151 | 9088057 | ||
BC151169.1 | - | 2 | 153 | 9088058 |

More Information
Other Species Targeted

Gene Ontology Categories:
