siRNA Details
Gene Symbol: Polr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081247.1 | NP_001074716.1 | 3 | 326 | ||
GenBank mRNA | AK039440.1 | - | 3 | 339 | ||
AK143315.1 | - | 3 | 276 | |||
AK170475.1 | - | 3 | 333 | |||
BC053071.1 | - | 3 | 289 | |||
BC151002.1 | - | 3 | 236 | 9087915 | ||
BC151168.1 | - | 3 | 234 | 9088057 | ||
BC151169.1 | - | 3 | 236 | 9088058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
