siRNA Details
Gene Symbol: Polr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081247.1 | NP_001074716.1 | 5 | 744 | ||
GenBank mRNA | BC053071.1 | - | 5 | 707 | ||
BC151002.1 | - | 5 | 654 | 9087915 | ||
BC151168.1 | - | 5 | 652 | 9088057 | ||
BC151169.1 | - | 5 | 654 | 9088058 |

More Information
Other Species Targeted

Gene Ontology Categories:
